Would you like to log in again?
Immunity Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T229-15 T229
470353-138EA 12.35 USD InStock
470353-138 470353-174
Immunity Microslides
Slides Prepared Slides
Investigate the cells of the immune system.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. Blood phagocyte (900×)/phagocytosis (2500×) 2. Monocyte (3000×)/macrophage (4000×) 3. Bone marrow (400×)/lymph gland (40×) 4. Thymus gland (175×) 5. Normal T-cells (2000×)/HIV budding from T-cell (2700×) 6. Plasma cell (37000×) 7. Normal red blood cells (3000×) - agglutinated cells (100×) 8. Eosinophil (1400×)/basophil (1400×).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty