Would you like to log in again?
The Kingdom Monera Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T213 T213-15
470353-192BX 179.4 USD InStock
470353-192 470353-180
The Kingdom Monera Microslides
Slides Prepared Slides
Learn about the Kingdom Monera.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects It on both sides
  • Set of 15 Includes detailed lesson pan on the topic under study
  • Durable, inexpensive teaching resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set Includes: 1. Staphylococcus aureus (1000×) 2. Bacillus subtilus (1000×) 3. Sirillium volutans (1000×) 4. Mycoplasma pneumonia (60,000×) 5. Rickettsia rickettsii (45,000×) 6. Cgleocapsa (1000×) 7. Oscillatoria (400×) 8. Nostoc - W.M. (100×) X.S. (400×).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty