Would you like to log in again?
PureAmp™ Color-Coded PCR Strips
PureAmp™
P3010-A-O P3010-A-P P3010-A-R P3010-A-B P3010-A-G P3010-A-Y
470353-210PK 93.5 USD InStock
470353-210 470353-212 470353-214 470353-206 470353-216 470353-218
PureAmp™ Color-Coded PCR Strips
Tubes PCR Tubes
Color-coded PCR strip tubes are easily identified at a glance, making them the ideal tube to track samples.
  • RNase, DNase, DNA, and PCR inhibitor free
  • 0.2 ml, or low profile 0.1 ml options
  • Ultra-thin, uniform wall thickness
  • White and frosted tubes for enhanced qPCR results
All PureAmp™ PCR tubes and strips are precision manufactured to maintain tight dimensional tolerances of ultra-thin wall thickness, for improved thermal transfer. PureAmp™ PCR plastics are certified free of RNase, DNase, DNA, and PCR inhibitors, and are made of heat resistant medical-grade polypropylene.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty