Would you like to log in again?
Large Volume Conical Centrifuge Tubes
C0225 C0175
470353-204CS 198.9 USD InStock
470353-204 470353-208
Large Volume Conical Centrifuge Tubes
Tubes Centrifuge Tubes
These polypropylene tubes are durable and centrifuge compatible.
  • Molded graduations
  • Leakproof screw caps
  • Sterile
  • RNase, DNase, and Pyrogen-free
Large volume conical centrifuge tubes offer a maximum RCF of 6000×g. These tubes may be used within a temperature range of –80 to 120 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty