Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Graduated Cylinders, Glass, Class A, Batch Certified, TD

Supplier: United Scientific Supplies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

UNBCCTD500 UNBCCTD2000 UNBCCTD250 UNBCCTD10 UNBCCTD50 UNBCCTD100 UNBCCTD25 UNBCCTD1000 UNBCCTD5
470353-244PR 30.8 USD InStock
470353-244 470353-266 470353-278 470353-248 470353-258 470353-228 470353-270 470353-294 470353-252
Graduated Cylinders, Glass, Class A, Batch Certified, TD
Cylinders Measuring Cylinders
Class A volumetric glassware provides the highest degree of accuracy.

  • Double metric scale, 'To Deliver'
  • Meets ASTM E1272, Class A standards, batch certified
  • One-piece molded glass construction with hexagonal base and adjustable plastic bumper guard
  • Made from heavy-duty borosilicate 3.3 glass
  • Strain point: 515 °C
  • Annealing point: 565 °C
  • Softening point: 820 °C

Graduated cylinders comply with ASTM E1272, Class A standards, batch certified. Each cylinder features a double metric scale, calibrated to deliver. Borosilicate glass 3.3 cylinders with pour spouts feature heavy uniform wall tubing and attached glass hexagonal bases. All cylinders include bumper guards and a batch calibration certificate.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty