Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
D Cell Battery Holder and Joiner
Supplier: United Scientific Supplies
BTHD01-PK/4
470353-350PK 5.72 USD InStock
470353-350
D Cell Battery Holder and Joiner
Batteries
Easily set up batteries in series or parallel.
  • Strong spring to hold batteries in place
  • Connects together in series or parallel
Single holders can be snapped together in parallel or series to extend capacity or voltage. Include male and female button snaps on ends to allow series connections. Also includes extensions on sides for parallel connections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty