Would you like to log in again?
AquaSprouts Fountains
Supplier: AQUASPROUTS, INC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AquaSprouts
6155 6148
470353-520EA 169.95 USD InStock
470353-520 470353-524
AquaSprouts Fountains
Educational Materials Biology Educational Materials Ecology Learning Activities
Versatile indoor or outdoor system which can be used for Aquaponics, Hydroponics, or Water Garden.
  • Rated for indoor and outdoor use
  • Durable, weatherproof grow bed and fountain reservoir
  • Includes a submersible 160 gph pump
  • Control the flow rate with customizable plumbing fittings
  • Room for 6 or more plants
Add a touch of beauty and a splash of life to any room with the AquaSprouts Fountain! Water feature, planter, and eye-catching decor, the Fountain can be used as an aquaponic or hydroponic system, or a simple water garden.
Similar to AquaSprouts Gardens, the expanded clay grow media replaces soil from traditional gardening and serves as the support for your plants.
Use any small freshwater fish to skip the added fertilizer and let Mother Nature be your guide. The fish provide nutrients for the plants and the plants clean the water for the fish. Once established this means a low maintenance ecosystem at your fingertips.
Not in the mood for fish? Use water-soluble nutrients to keep your plants happy and healthy with ease. Enjoy a tranquil water garden while growing your own personal oasis.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty