Would you like to log in again?
Test Tube Racks for 25 mm Tubes
Supplier: United Scientific Supplies
77804 77809
470353-632EA 20.99 USD InStock
470353-632 470353-646
Test Tube Racks for 25 mm Tubes
Racks Tube Racks
Polycarbonate racks provide clear views to tubes.
  • 3-tier design
  • Transparent plates for clear view of tubes
  • Holds tubes up to 25 mm in Ø
These polycarbonate racks with a 3-tier design provide clear view of the tube contents and convenient insertion and removal of tubes. Upper and middle plates are transparent. Beveled holes on bottom plate provide stability to standing tubes. Racks are autoclavable and can withstand sub-freezing temperatures. Available in 12-tube and 18-tube configurations, each able to hold tubes up to 25 mm in Ø.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty