Would you like to log in again?
Screen Sieve Set 6
Supplier: GSC INTERNATIONAL SE
SSC-15-60 SSC-15-230 SS-15-5 SSC-15-10 SSC-15-35 SS-15-6 SSC-15-120
470353-760EA 14.5 USD InStock
470353-760 470353-762 470353-764 470353-766 470353-758 470353-768 470353-770
Screen Sieve Set 6
Sieves Test Sieves
  • Set includes 6 popular mesh sizes
  • The frame is made from durable steel
  • A top and bottom cover are provided for storage
  • Each screan is labeled with the mesh size
The process of screening allows students to study the make up of the materials they have collected. The screening process athat aid in the recognition of the different parts of a specimen. The screen sieves are a valuable tool used in this process. This set features steel screens with six different sizes of mess that include: 5, 10, 35, 60, 120, 230. A top and bottom is also provided to enclose the set.
Ordering information: Each screen can be purchased individual.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty