Would you like to log in again?
CENCO® Engraved Precision Force Table
Supplier: VWR International
CENCO®
PH0347AVWR
470353-930EA 850 USD InStock
470353-930
CENCO® Engraved Precision Force Table
Educational Materials Physics Educational Materials Mechanics Learning Activities
Rugged metal construction and innovative features make this tool an industry standard.
  • Larger center ring, with printed bullseye target make it easy to read
  • Four low-friction pulleys with stainless steel hanging mass sets
  • Four separate nylon pulley cords with quick release clips and rings
  • Assortment of additional slotted masses included
  • Comprehensive teacher guide included
The CENCO® Precision Force Table is an elegant tool for studying static equilibrium and vector addition. The 40 cm aluminum tabletop is engraved with a 360° graduated scale. The pulley clamps are specially designed to conform to the curve of the circular table, so the pulleys will not wobble. These low-inertia pulleys with miniature bearings ensure parallax-free angle readings. The sturdy support rod and heavy tripod base with leveling screws give the table stable support, and the heavy center pin fits into the tabletop for easy assembly. Quick-release clips allow users to easily switch between 2, 3, or 4 applied forces. Additional small slotted masses make this a truly precision apparatus. This fine piece of equipment is packed with useful features, and will last for many years in your classroom.
Accessories information: A replacement ring and string for the 40 cm force table is also available and sold separately. Fits force tables with 40 cm diameter. Pre-assembled top ring with four lines. Rings attached on each line for hanging masses. Includes one replacement part for the 40 cm diameter table top. The part consists of one top ring for above the table and four 12" strings with rings for the four pulleys.
Delivery information: The slotted masses assortment includes four 50 g, four 100 g, two 10 g, and two 20 g.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty