Would you like to log in again?
Supertek® Dual Scale Spring Scales
Supplier: Shiv Dial Sud & Sons
Supertek®
PH10132/7 PH10132/6 PH10132/3 PH10132/1 PH10132/4 PH10132/2 PH10132/5
470353-980EA 10 USD InStock
470353-980 470353-982 470353-996 470353-998 470353-986 470353-978 470353-990
Supertek® Dual Scale Spring Scales
Balances and Scales
Plastic spring scales, built to last in any laboratory.
  • Scale calibrated in both grams and Newtons
  • Built-in zero adjustment
  • Crystal clear plastic body for easy viewing
These reliable spring scales are housed in a robust plastic transparent body for easy viewing. Overall length of approximately 285 mm, with upper and lower suspension hooks and zero adjustment. Scales are calibrated in both grams and Newtons.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty