Would you like to log in again?
Raspberry Pi 4 Starter Kits
Supplier: NEWARK ELECTRONICS
80AH9362 80AH9359
470354-012KT 106.29 USD InStock
470354-012 470354-000
Raspberry Pi 4 Starter Kits
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Complete Raspberry Pi kit contains everything needed to get started in single board computing.
  • Perfect for makerspaces and Fab labs, as well as computer science classrooms
  • Includes a 16 GB microSD card loaded with Raspberry Pi OS for easy start up
  • Included 3 A power adapter provides enough power for advanced applications
  • Includes 3' micro HDMI to HDMI cable for simple connections to an existing display
Introducing the Raspberry Pi 4 Model B, with 4 GB RAM. At its foundation, this R-Pi offers increased features and capabilities, while maintaining full backwards compatibility with the earlier Raspberry Pi 3 Model B+. This product’s key features include a high-performance 64-bit quad-core processor, dual-display support at resolutions up to 4K via a pair of micro-HDMI ports, hardware video decode at up to 4Kp60, 4 GB of RAM, dual-band 2.4/5.0 GHz wireless LAN, Bluetooth 5.0, Gigabit Ethernet, USB 3.0, and PoE capability (via a separate PoE HAT add-on). The dual-band wireless LAN and Bluetooth have modular compliance certification, allowing the board to be designed into end products with significantly reduced compliance testing, improving both cost and time to market.
Delivery information: Kit includes: Raspberry Pi 4 model B, with 4 GB RAM, 5.1 V, 3 A power supply, 16 GB Micro SD card, preloaded with NOOBS operating system, 1M Micro HDMI to HDMI cable, two-piece heat sink kit, molded enclosure (white), getting started booklet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty