Would you like to log in again?
3Doodler Pro+ 3D Printing Pen Set
Supplier: D&H
3Doodler Pro+
3DP2BKALL
470354-042EA 234.99 USD InStock
470354-042
3Doodler Pro+ 3D Printing Pen Set
Printers 3D printers
A professional-grade 3D printing solution.
  • Robust dual-drive system built to run longer and harder than any other 3D pen
  • Versatile enough to accept ABS, PLA, Metal, FLEXY, and Nylon filaments
  • Plug-in power supply and temperature selection options ensure the Pro+ pen operates at the optimal temperature
The 3Doodler Pro+ combines sleek, ergonomic styling with advanced technology for superior performance to elevate design and bring ideas to life. Explore 3D design and prototypes in a fraction of the time and share ideas using a range of materials from ABS to metal and nylon — the possibilities are endless.
The PRO+ rests comfortably in the hand and features a magnetic maintenance panel that enables easy access the pen’s interior. The digitally adjustable speed and temperature settings offer unparalleled design control. Create professional-grade Doodles with 6 packs of refills, including ABS, PLA, Metal, Flexy, and Nylon, equivalent to 125 feet of plastic and over 1,500 feet of extrusion.
There is also a Nozzle Set included which enables you to swap out between six different nozzle tips to customize your outcome. It includes a ribbon tip, square tip, triangle tip, a super fine 0.5 mm nozzle, and thicker 1 mm and 1.5 mm nozzles.
Delivery information: Includes (1) Pro+ 3D printing pen; (1) nozzle set; (1) Mini DoodlePad; (1) unblocking tool; (6) refill packs - including ABS, PLA, metal, Flexy, and Nylon; detailed product manual and quick start guide; (1) set of international adapters.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty