Would you like to log in again?
United Scientific Economy Hall's Carriage
Supplier: United Scientific Supplies
United Scientific
HLCRG1-E
470354-332CS 406.75 USD InStock
470354-332
United Scientific Economy Hall's Carriage
Educational Materials Physics Educational Materials Mechanics Learning Activities
Smoth rolling hall's carriage for velocity experiments.
  • Smooth rolling wheels
  • Well for holding masses
  • Includes hole for attaching string
This economy Hall’s carriage is perfect for demonstrations and investigating principles such as Newton’s laws, acceleration, velocity and friction. This economy Hall’s carriage has a durable plastic body with a roomy well for additional masses (not included). The hole on the front bumper allows you to easily connect a string for pulling the cart up a ramp. Both the body and wheels are made from high impact plastic.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty