Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Solderless Breadboard One Column

Supplier: United Scientific Supplies
SLSBRBD01
470354-334EA 12.1 USD InStock
470354-334
Solderless Breadboard One Column
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Breadboard perfect for smaller electronic projects.

  • Accepts most electronic components
  • Use to debug circuit designs
  • Breadboard strips connect together to form larger working areas
  • Pins are identified by numbered rows and columns

The 630-point solderless breadboard is built with high-quality construction that can withstand repeated use. It includes color coordinates that are screen-printed for easy reference. This breadboard is perfect for investigating circuit designs and prototyping. It accepts most electronic components including integrated circuits and transistors. The breadboard comes with three binding posts and four rubber feet.

Specifications:
Bus strips: 2
Contact points: 630
0.1" hole spacing
Breadboard size: 6.5L×2.3" W
Baseplate size: 7.25L×4.25" W
Weight: 0.50 lbs.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty