Would you like to log in again?
Eisco Heavy Wall Test Tubes
Supplier: EISCO SCIENTIFIC LLC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Eisco
CH0719A CH0719F CH0719E CH0719D CH0719C CH0719B
470354-592PK 93.6 USD InStock
470354-592 470354-590 470354-588 470354-598 470354-596 470354-594
Eisco Heavy Wall Test Tubes
Tubes Test Tubes
Durable heavy wall test tubes.
  • Borosilicate glass 3.3, heavy wall, with rim
  • Strong resistance to heat and chemicals
  • 1.5 mm wall thickness
These products have a heavy wall with a rim. Has strong resistance to heat and chemicals. Comes in bulk packs. Comes in multiple different capacities.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty