Would you like to log in again?
Innovating Science Unknown Compositions Samples: Soda Ash
Supplier: ALDON CORP SE
Innovating Science
IS5902
470354-896EA 40 USD InStock
470354-896
Innovating Science Unknown Compositions Samples: Soda Ash
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
A series of sodium carbonate (soda ash) samples with a range of known purities.
  • 10×100 g samples with purity concentrations ranging from 20 to 50%.
  • Reference sheet included
Use these in practicing analytical techniques such as gravimetry and acid-base titration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty