Would you like to log in again?
Innovating Science Unknown Compositions Samples: Ferrous Ammonium Sulfate
Supplier: ALDON CORP SE
Innovating Science
IS5901
470354-898EA 144.5 USD InStock
470354-898
Innovating Science Unknown Compositions Samples: Ferrous Ammonium Sulfate
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
A series of ferrous ammonium sulfate samples with a range of known purities.
  • 20×100 g samples with purity concentrations ranging from 0.1 to 13% iron
  • Reference sheet included
Use these in practicing analytical techniques such as gravimetric, potentiometric, and spectrophotometric analyses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty