Would you like to log in again?
Wallcur® Transofix Transfer Spike
Supplier: WALLCUR, LLC.
Wallcur®
223TS
470354-994BG 65.4 USD InStock
470354-994
Wallcur® Transofix Transfer Spike
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Fast and easy transfer.
  • Allows excellent flow even with non-evacuated glass vials
  • Fits into rubbers stoppers 6.3 mm and bag set ports
  • Large internal diameter
  • Provides secure connection of two containers
  • Large grip flanges
Transofix is a needle-less double transfer spike made by braun for rapid transfer of medication or diluent between rubber stoppered containers. This needle-less transfer system allows for the transfer of a diluent into powdered medication without the need for repeated piercing of vials. Ideal for use with Wallcur's practi-Powder® vials or Wallcur's practi-alteplase powder and diluent vials. This simulation is identical to the real clinical experience. Affordable, dependable, and reusable.
Delivery information: These products require a release form for purchase. If your account does not have one on file, the item cannot ship until release documentation is received.
Caution: For training purposes only. Not for human or animal use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty