Would you like to log in again?
PicoSolutions Solar Racer Plus Kit
Supplier: PICOSOLUTIONS GROUP INC SE
PS-3027-03-D4 PS-3027-03-D3 PS-3027-03 PS-3027-03-D2 PS-3027-03-D1 PS-3027-03-PK
470355-380EA 2214.15 USD InStock
470355-380 470355-382 470355-304 470355-348 470355-366 470355-346
PicoSolutions Solar Racer Plus Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
DIY solar car kit: Build and learn solar-powered mechanics.
  • Suitable for ages 7+
  • Understand solar energy by building and optimizing a solar-powered card
  • 15 minute assembly with without soldering or batteries
  • Customizable with free 3D print designs
Build and test a solar car the solar racer 2.0 is a fun and educational STEM tool for K12 classrooms, camps, after-school programs, or learning at home. It helps students learn about solar energy while they enjoy making the car faster.
Compared to the standard version (470355-322) the plus version includes options for both 1.5 and 7.2 V solar panels and an extra motor for increased power, making your racer even faster.
Ordering information: DESS course for advanced learning: An upgrade to the DESS course enriches STEM education, focusing on design, entrepreneurship, and innovation, aimed at honing problem-solving skills. Prepares students for future: By blending fun with education, the solar racer 2.0 and DESS course prepare students for a tech-focused future, equipping them to address real-world challenges.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty