Would you like to log in again?
PicoSolutions Horizontal Wind Energy Standard
Supplier: PICOSOLUTIONS GROUP INC SE
PS-3001-01-PK PS-3001-01 PS-3001-01-D2 PS-3001-01-D3 PS-3001-01-D1 PS-3001-01-D4
470355-350EA 306.46 USD InStock
470355-350 470355-360 470355-318 470355-300 470355-340 470355-376
PicoSolutions Horizontal Wind Energy Standard
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Explore horizontal wind energy with a DIY hands-on turbine model.
  • 6th grade to college level tailored activities for different learning stages
  • Subjects spanning sustainable energy, math, aerodynamics, geometry
  • Easy 10 minute assembly
Build and test a wind turbine to teach the power of the wind features a DC motor for beginners and an AC generator with 12+ experiments for advanced learners. The kit uses an LED, a buzzer, or a multimeter to show how wind generates electricity. The kit includes all parts for building the turbine, with blade adapters for custom blades made from dowels or 3D printing.
Ordering information: Educational support bundle: Upgrade available for enhanced learning with support from a certified virtual teacher mentor and DESS course. Future-ready skills: The DESS course enriches the kit by integrating design, entrepreneurship, and STEM, fostering creativity, innovation, and problem-solving abilities.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty