Would you like to log in again?
PicoSolutions Vertical Wind Turbine Standard Kit
Supplier: PICOSOLUTIONS GROUP INC SE
PS-3014-01 PS-3014-01-D4 PS-3014-01-D2 PS-3014-01-D1 PS-3014-01-PK PS-3014-01-D3
470355-316EA 1844.92 USD InStock
470355-316 470355-312 470355-334 470355-384 470355-320 470355-342
PicoSolutions Vertical Wind Turbine Standard Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Explore vertical wind energy with a DIY hands-on tubine model.
  • 6th grade to college level tailored activities for different learning stages
  • Subjects spanning Sustainable energy, math, aerodynamics, geometry
  • Easy 10 minute assembly
Build and test a wind turbine to teach the power of the wind the basic model has a DC motor generator and uses an LED, a buzzer, or a multimeter to show how wind generates electricity. The kit includes all parts for building the turbine.
Ordering information: Enhanced learning option: Upgrade available for a DESS course to integrate design, entrepreneurship, and STEM skills, fostering innovation and problem-solving abilities.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty