Would you like to log in again?
PicoSolutions Vertical Wind Turbine Plus Kit
Supplier: PICOSOLUTIONS GROUP INC SE
PS-3014-02 PS-3014-02-D4 PS-3014-02-PK PS-3014-02-D2 PS-3014-02-D1 PS-3014-02-D3
470355-378EA 244.92 USD InStock
470355-378 470355-356 470355-314 470355-324 470355-352 470355-362
PicoSolutions Vertical Wind Turbine Plus Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Explore vertical wind energy with a DIY hands-on tubine model.
  • 6th grade to college level tailored activities for different learning stages
  • Subjects spanning sustainable energy, math, aerodynamics, geometry
  • Easy 10 minute assembly
Build and test a wind turbine to teach the power of the wind the kit students to experiment with wind power using DC and AC configurations and showcasing the power with an LED, a piezo buzzer, or a multimeter. The kit uses an LED, a buzzer, or a multimeter to show how wind generates electricity. The kit includes all parts for building the turbine.
Compared to the standard model (470355-312), the plus version includes the AC generator with magnets, coils, and over 12 electricity labs.
Ordering information: DESS course enhancement: Offers an upgrade to a specialized course that melds design, entrepreneurship, and STEM, aimed at cultivating creativity, innovation, and problem-solving prowess.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty