Would you like to log in again?
PicoSolutions Alternative Energy Education Lab Standard Kit
Supplier: PICOSOLUTIONS GROUP INC SE
PS-3019-01-D3 PS-3019-01-PK PS-3019-01-D1 PS-3019-01-D2 PS-3019-01 PS-3019-01-D4
470355-308PK 1169.07 USD InStock
470355-308 470355-372 470355-336 470355-302 470355-330 470355-332
PicoSolutions Alternative Energy Education Lab Standard Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Explore alternative energy with a DIY hands-on turbine model.
  • 3rd grade to college level tailored activities for different learning stages
  • Includes DC turbines (horizontal and vertical) and a 7.2 V solar panel for a full spectrum of alternative energy exploration
  • Versatile configuration Supporte enables both AC and DC configurations
  • Subjects spanning sustainable energy, math, aerodynamics, geometry
Build and test a wind turbine kit contents includes vertical and horizontal turbine level 1 kits, six hydro paddles, a multimeter, and a 7.2 V 200 mA solar panel.
Ordering information: DESS course upgrade: Enhance learning with an upgrade to our design entrepreneurship and STEM skills (DESS) course, designed to cultivate creativity, innovation, and problem-solving skills, preparing students for a future in technology and engineering.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty