Would you like to log in again?
PicoSolutions Horizontal Wind Energy Plus Kit
Supplier: PICOSOLUTIONS GROUP INC SE
PS-3001-02-D2 PS-3001-02-D4 PS-3001-02 PS-3001-02-D3 PS-3001-02-D1 PS-3001-02-PK
470355-392EA 3075.68 USD InStock
470355-392 470355-338 470355-306 470355-344 470355-298 470355-386
PicoSolutions Horizontal Wind Energy Plus Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Explore horizontal wind energy with a DIY hands-on turbine model, now including AC and DC power generation.
  • 3rd grade to college level tailored activities for different learning stages
  • Subjects spanning sustainable energy, math, aerodynamics, geometry
  • Easy 10 minute assembly
Build and test a wind turbine to teach the power of the wind Features a DC motor for beginners and an AC generator with 12+ experiments for advanced learners. The kit uses an LED, a buzzer, or a multimeter to show how wind generates electricity. The kit includes all parts for building the turbine, with blade adapters for custom blades made from dowels or 3D printing.
In comparsion to the standard kit (470355-376), the plus kit includes an AC generator with 12+ experiments for advanced learners.
Ordering information: Educational support bundle: Upgrade available for enhanced learning with support from a certified virtual teacher mentor and DESS course.
Future-ready skills: The DESS course enriches the kit by integrating design, entrepreneurship, and STEM, fostering creativity, innovation, and problem-solving abilities.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty