Would you like to log in again?
American Scientific Magnetic Iron Filings Chips
Supplier: AMERICAN SCIENTIFIC LLC SE
American Scientific
3124-00
470355-436EA 12 USD InStock
470355-436
American Scientific Magnetic Iron Filings Chips
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Pieces of reusable zinc-plated iron wire for magnetic field experiments.
  • Magnetic properties for suitable for demonstrating magnetic fields
  • Fine and easily manipulable texture, but much easier to clean up than iron filings
  • Safe and non-toxic material
  • 50 g packaged in a reusable storage vial
All of the uses of iron filings for experiments with magnetic fields, with none of the mess! These pieces of reusable zinc-plated iron wire (26 to 27 AWG) replace the messy, rust-prone iron powder typically employed in demonstrations of magnetic fields. The short pieces of wire align themselves linearly to provide an excellent representation of magnetic lines of force. Use them to your advantage in any demonstration requiring clear, curvilinear force lines. Clean up is a snap - no more iron dust permanently clinging to your magnets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty