Would you like to log in again?
Grain of Wheat Bulbs
Supplier: AMERICAN SCIENTIFIC LLC SE
9125-12 9125-03
470355-458PK 130 USD InStock
470355-458 470355-422
Grain of Wheat Bulbs
Lights Incandescent Lights, Lamps and Bulbs
Flexible and easy alternative to standard mini bulbs: no holder required!
  • Comes with a teacher’s guide
  • Sold in packs of 100
  • Incandescent bulbs take a maximum of 12 V and 500 mA
  • LED bulbs take a maxiumum of 3 V and 3 mA
This is a new generation of ultra-bright bulbs offering easy replacement, making them ideal for use in a variety of miniature equipment and circuit applications. The wheat bulbs can also be used in control panels and high-quality commercial and industrial controls. They are similar to the wire-ended lamps but they have stranded wire leads about 8" (200 mm) long. The bulb is about 0.13" (3 mm) dia. and 0.25" (6 mm) long - the size of a grain of wheat.
Or, upgrade your outdated bulbs to the 21st century! These white color LED bulbs offer an easy replacement, making them ideal for use in a variety of miniature equipment and circuit applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty