Would you like to log in again?
PicoSolutions Robotic Hand Standard Kit
Supplier: PICOSOLUTIONS GROUP INC SE
PicoSolutions
PS-3020-01-D2 PS-3020-01-D1 PS-3020-PK PS-3020-01-D3 PS-3020
470355-746EA 400 USD InStock
470355-746 470355-744 470355-756 470355-754 470355-752
PicoSolutions Robotic Hand Standard Kit
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Codeable, 3D-printed robotic hand for interactive STEM learning.
  • Educational focus on medical innovation
  • Easy to use and assemble: features a solderless breadboard and simple components
  • Powered by Arduino Uno
  • Optional online courses foster creativity and problem-solving in a tech-focused educational environment
The robotics hand standard kit is perfect for learning about medical innovation and bionics. It includes everything needed to build a robotic hand: a 3D-printed hand model, five micro servo motors to move the hand, servo caps for connecting the motors, hookup wires for connections, an Arduino Uno for control, a USB cable for connecting to a computer, an extra USB wire, two types of capacitors for circuit stability, a voltage regulator, and a solderless breadboard for easy circuit building. This kit is great for students and hobbyists interested in robotics and medical technology.
Upgrade to the special Design Entrepreneurship and STEM Skills (DESS) courses, with extra support. These courses combine design, entrepreneurship, and STEM education to boost creativity and innovation. It's designed to develop top-notch problem-solving skills, preparing students for a tech-focused future and enabling them to tackle complex challenges effectively.
Delivery information: Kit Includes a robotic hand model, five micro servo motors, servo caps, hookup wires, and more for a complete assembly experience.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty