Would you like to log in again?
PicoSolutions 4-Axis Robotic Arm
Supplier: PICOSOLUTIONS GROUP INC SE
PicoSolutions
PS-4000-01 PS-4000-01-PK PS-4000-01-D2 PS-4000-01-D1 PS-4000-01-D3
470355-748EA 1500 USD InStock
470355-748 470355-758 470355-742 470355-740 470355-750
PicoSolutions 4-Axis Robotic Arm
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
4-axis educational robotic arm: precision learning tool.
  • 4 precision axes: offers a broad range of motion for versatile tasks from simple placements to complex maneuvers
  • User-friendly coding: works seamlessly with major block-based coding platforms like Scratch and Blockly
  • Compact design is ideal for desktop use in classrooms, homes, or labs; maximize space without sacrificing functionality
  • Easily customizable with add-ons like sensors and attachments to grow with user skills and demands
  • Supplied with a wealth of tutorials and learning materials to support users of all experience levels
Introducing the mini educational 4-axis robotic arm – a gateway to the world of robotics and coding. This compact yet powerful robotic arm is designed to teach users of all ages about the fundamentals of robotics, programming, and automation. Standing as a marvel of modern engineering, the Mini 4-Axis Robotic Arm boasts four precision-controlled axes, allowing for a wide range of movement and functionality. From simple pick-and-place tasks to more complex manipulations, this versatile arm can handle it all with ease.
What truly sets this robotic arm apart is its intuitive block coding interface. No prior coding experience is required – simply drag and drop blocks of code to command the arm's movements and actions. With a user-friendly interface, beginners can quickly grasp the basics of programming while seasoned enthusiasts can delve into more advanced concepts. Whether you're a student exploring STEM subjects, an educator looking to enhance classroom learning, or a hobbyist eager to dive into robotics, the Mini Educational 4-axis robotic arm offers endless possibilities for experimentation and discovery.
Upgrade to the special Design Entrepreneurship and STEM Skills (DESS) courses, with extra support. These courses combines design, entrepreneurship, and STEM education to boost creativity and innovation. It's designed to develop top-notch problem-solving skills, preparing students for a tech-focused future and enabling them to tackle complex challenges effectively.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty