Would you like to log in again?
United Scientific™ Flat Electrode Strips
Supplier: United Scientific Supplies
United Scientific™
UNELCCU UNELCTIN UNELCFE UNELCSET UNELCAL UNELCZN UNELCC
470356-060EA 2.77 USD InStock
470356-060 470356-030 470356-068 470356-014 470356-058 470356-020 470356-054
United Scientific™ Flat Electrode Strips
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A variety of electrodes for electrochemical cells.
  • Available in six different metals
Use these flat, rigid electrodes in a variety of electrochemical demonstrations and experiments. The 127 mm long strips can fit into a variety of beakers, wet cells, and porous cups. If needed, the top can be bent over to stand upright in a container. Since the strips consist of one type of metal, they can be used in electroplating demonstrations. Also available as a set of 12 electrodes (two each of aluminum, carbon, copper, iron, tin, and zinc).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty