Would you like to log in again?
Earthquake Shake Table
Supplier: United Scientific Supplies
ERTHQUAKSIM
470356-072KT 85.8 USD InStock
470356-072
Earthquake Shake Table
Educational Classroom Kits and Activities
Effectively test your earthquake-proof building.
  • Investigate horizontal and vertical vibration
  • Motors provide reliable and consistent motion for demonstration
  • Broad base for a variety of building designs
  • Great for tabletop demonstrations or small group learning
  • Activity guide included
This Earthquake Shake Table allows students to test different structures to see which ones can best withstand the shaking caused by earthquakes. The base has four strong springs at the corners with adjustable mountings to hold a variety of tower bases. A tower model can have a base up to 12" on each side. A pair of motors are mounted underneath and have a protective cage to protect fingers while the table is in motion. The motors can be easily powered powered individually or together by a 1 to 5 V DC power supply (required but not included), and are intended to be a reliable and consistent source of vibrations. If a more energetic vibration is needed to test a design, the base can be manually shaken.
Ordering information: This is a shaking apparatus only. Users need to provide the building materials and power supply. Power supply available for separate purchase.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty