Would you like to log in again?
PinPoint™ Pipette Controller
Supplier: United Scientific Supplies
PinPoint™
UNPINPNT04 UNPINPNT05 UNPINPNT01 UNPINPNT03 UNPINPNT02
470356-094EA 5.94 USD InStock
470356-094 470356-090 470356-092 470356-086 470356-088
PinPoint™ Pipette Controller
Pipettes Pipette Controllers
Digital display makes transferring liquids easier.
  • Compatible with glass or plastic serological pipettes (0.1 to 100 ml)
  • LCD indicates battery charge and 6 speed levels
  • Fills a 25 ml pipette in less than 5 seconds
  • Lithium-ion battery charges fully within 3 hours and can provide up to 8 hours of use
Ultra-light and powerful pipette controller is designed for comfort as well as accuracy. Intuitive 6 level speed setting and sensitive triggers provide a wide range of aspirating/dispensing speeds. Pipette controller is ergonomically designed for extended usage.
Certifications: Meets the requirements of the applicable EU directives, UK regulations, UL, and CAN/CSA.
Ordering information: The PinPoint™ Pipette Controller comes with an AC adapter with USB connection, a 0.45 μm hydrophobic filter, a user manual, wall mount, and two-position desk stand.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty