Would you like to log in again?
OHAUS® Laboratory Vortex Mixers
Supplier: OHAUS CORP.
30392115 30392122 30392108 30392129
470356-208EA 345 USD InStock
470356-208 470356-206 470356-202 470356-198
OHAUS® Laboratory Vortex Mixers
Shakers and Mixers Vortex Mixers
Offering gentle to high-speed reliable mixing.
  • Touch mode operation activates mixing when depressing the cup head or continuous mode for use with accessory attachments
  • Robust digital and non-digital vortex mixers offer effective speed control from low RPM start-up for gentle shaking to high-speed mixing for vigorous vortexing of samples
  • Sturdy design stabilizes mixer at all speeds
  • The fixed speed vortex mixer is an economical choice for vigorous mixing of samples at high speed. To ease repetitive sample processing, this unit provides simple, effective full-speed mixing
OHAUS vortex mixer models allow gentle to high-speed reliable mixing. The fixed speed mixer is an economical choice for vigorous mixing of samples at high speed. The analog model offers variable speed control, and the digital model allows input of the exact speed and time to achieve repeatable results every time. The pulsing model features a unique pulse action that reduces heat generation, while providing more effective mixing and cell disruption.
Certifications: Product Safety: CAN/CSA C22.2 61010-1, CAN/CSA C22.2 61010-2-051, UL 61010-1
Electromagnetic Compatibility: ICES-003 Class A, FCC Part 15 Class A
Compliance Marks: TÜV SÜD
Caution: https://us.ohaus.com/en-us/legal/caprop65
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty