Would you like to log in again?
OHAUS® Endeavor™ Open Air Orbital Shakers
Supplier: OHAUS CORP.
Endeavor™
30770909 30770910
470356-204EA 1300 USD InStock
470356-204 470356-210
OHAUS® Endeavor™ Open Air Orbital Shakers
Shakers and Mixers Orbital Shakers
Stable, long-term shaking with triple eccentric drive and maintenance-free brushless DC motor.
  • Adaptable to a wide variety of accessories to customize for low to mid volume applications
  • Programmable with a simple on-board menu to save time and guarantee reproducible results
  • Remote function permits PC control and data transfer
For use in a wide variety of applications including Microbiology, Molecular Biology, Cell Cultures, Staining/Destaining, DNA Studies, Environmental and Food Sciences, Chemistry, General Mixing and more.
Feature a bright, backlit LCD display for speed and time. The timer will count up elapsed time or when set, timer will count down and shut off shaking when time reaches zero. A green indicator light illuminates when the unit is shaking with visual and audio alarm when timer counts down to zero or an error occurs.
Small footprint to save bench space, durable chemical-resistant polymer housing, simple knob control for timer and shaking, easy to navigate menu for advanced features.
Certifications: Product Safety: CE, TUV, TÜV SÜD certified to CAN/CSA C22.2 61010-1 ; CAN/CSA C22.2 61010-2-051; UL 61010-1; UL 61010-2-051.
Electromagnetic Compatibility: Canada ICES-003 Class B; FCC Part 15 Class B.
Delivery information: Supplied with a sturdy, easy-to-clean aluminum tray with attachment points for flask clamps, tube racks and non-slip rubber mat. Optional accessories include universal platform and over 20 flask clamps and tube racks.
Caution: https://us.ohaus.com/en-us/legal/caprop65
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty