Would you like to log in again?
Eisco Rectangular Retort Stand, Rod, Clamp and Ring Set
Supplier: EISCO SCIENTIFIC LLC
Eisco
MTST1SPL
470356-224EA 33 USD InStock
470356-224
Eisco Rectangular Retort Stand, Rod, Clamp and Ring Set
Support Stands
The most compact retort stand, rod, clamp and ring set on the market!
  • Durable construction
  • Four piece set
This comprehensive set now features a two-piece stainless steel rod that effortlessly screws together, ensuring easy setup and enabling more compact storage when not in use. The retort stand is made of corrosion and scaling resistant, powder coated pressed steel, and features a tapped hole, suitable for connecting the included rod, or any other rods with a 10×1.5 mm metric thread. The stand measures 8" long, 5" wide and is approximately 1" in height. Included rod is made of stainless steel and measures 23.6" tall, 10 mm in diameter and features a 10×1.5 mm metric thread. The ring features a zinc plated steel shank secured to a screw clamp. Distance from each of the rings centers to the center of the support measures 4.75". Clamp is four-pronged with a cork lining.
Ordering information: The kit includes retort base, two piece stainless steel rod, 3" support ring, and a four-pronged cork-lined clamp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty