Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Synthetic Owl Pellets

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

OWLPELLET03 OWLPELLET01 OWLPELLET02
470356-226EA 16.5 USD InStock
470356-226 470356-238 470356-234
Synthetic Owl Pellets
Educational Materials Biology Educational Materials Ecology Learning Activities
Synthetic owl pellets provide a safe, realistic, and reusable dissection experience, featuring hand-crafted replicas of skeletons and carefully imitated contents that closely mimic natural owl pellets.

  • A clean and safe dissection experience
  • Realistic replica of natural owl pellet
  • Handmade
  • Reusable

Our simulated Owl pellets offer a hands-on approach to learning the intricacies of food webs and ecosystems through safe, realistic, and engaging replicas. Perfect for schools and colleges looking to enrich their biology or ecology curriculum, these simulated pellets provide a unique insight into the food chain and predator-prey relationships without the mess or safety concerns of real owl pellets. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.

Dimensions: 3×1.5×1.5"
Weight: 0.2 lbs.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty