Would you like to log in again?
Digital Force Meter
Supplier: GSC INTERNATIONAL SE
N-00001 N-00001-10
470356-480EA 84.08 USD InStock
470356-480 470356-460
Digital Force Meter
Dynamometers
Ideal for addition and resolution of forces, Newton's laws, and statics experiments.
  • Wide measureable range and decimal resolution makes this instrument a versatile alternative to traditional spring scales
With a wide range of 2, 5, 10 and 20 Newtons, this force meter replaces four types of spring balances. With the hold function, you can retain the measured value for added convenience. With the one-touch zero-calibration function, you can measure in any direction, 360 degrees, for complete accuracy. Compact and convenient, this digital force meter measures 157 mm in length, 45 mm in width, and 24 mm in height, making it easy to use and store.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty