Would you like to log in again?
HyFlex® 11-100 Static Control Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyFlex®
205592 205591 205593 205590 205589
80092-584CS 2354.41 USD InStock
80092-584 47727-530 47727-526 47727-528 47727-524
HyFlex® 11-100 Static Control Gloves, Ansell
Gloves
Designed for micro-electronic work where electro-static discharge (ESD) must be avoided, these comfortable industrial gloves with nitrile foam ensure that surfaces stay clean and uncontaminated. X-Static® yarn and static-reducing coating help prevent damage to sensitive electronic components and injury.
  • Ensure uncontaminated surfaces
  • Static-reducing coating
  • Oil is channeled away for a sure grip
  • Tolerance: ±0.5"
A thin design improves the handling of ever smaller components, while the ergonomic liner allows for precision in assembly applications.
Use in the aerospace, appliance manufacturing, electronics manufacturing, medical laboratory and research laboratory industries. Uses include assembling; inspecting and packing small or delicate parts; electrical component assembly and installation.
The foam nitrile-coated palm channels oil away from the glove surface for a sure grip, enabling detailed handling of small parts.
Certifications: Antistatic according to EN1149. Produced in ISO 9002-registered manufacturing facilities.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty