Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MiniSpin® Personal Microcentrifuges, 230 V

Supplier: Eppendorf
MiniSpin®
022620151
47727-634EA 2340.66 USD InStock
47727-634
MiniSpin® Personal Microcentrifuges, 230 V
Centrifuges Microcentrifuges
Microcentrifuges are compact and easy to use, yet powerful enough to enable each workstation to be equipped with a 'personal' centrifuge for a wide range of molecular biology separations and quick spins

  • 230 V model

Both are extremely compact with an exceptionally small footprint, which saves valuable space at crowded workstations. Both models have a remarkably quiet operation and a quick and smooth acceleration of <13 seconds and a deceleration of <12 seconds. They also feature a maintenance-free drive, a separate 'Short Spin' key for fast, convenient spins, and a lid that opens automatically at the end of the run, reducing handling time. Excellent air flow minimizes heating, protecting temperature-sensitive samples.

The MiniSpin® plus features a clear, easy-to-read digitial display of time and speed, as well as digital set in rpm or RCF. A rotor is included.

Two models are available in 230 V, 50 Hz to meet your requirements.

Caution: 230 V model. Corresponding models with voltage suitable for use in US (47727-632 and 47727-636) are also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty