Would you like to log in again?
MiniSpin® MiniSpin®/MiniSpin® plus
Supplier: Eppendorf
022620207 022620258 022620100 022620100 022620207
47727-636EA 2871.03 USD InStock
47727-636 CA47727-636 47727-638 47727-632 CA47727-632
MiniSpin® MiniSpin®/MiniSpin® plus
Centrifuges Microcentrifuges
Eppendorf MiniSpin® and MiniSpin® plus centrifuges are powerful, entry-level, easy-to-use centrifuges that are small enough to allow individual workstations to be equipped with their own. Key parts are made of metal for maximum safety, and the MiniSpin® plus is fast enough for molecular biology separations.
  • Extremely compact footprint
  • Acceleration and deceleration time <13 s
  • User-friendly digital display for time and speed
  • Metal rotor bowl; Rotors available: 2
  • Separate short-spin key for fast and comfortable centrifugation
  • Optimal ventilation flow reduces heating and protects temperature-sensitive samples
  • Soft-touch lid closure for ergonomic lid locking
  • Automatic lid opening at the end of the run to prevent sample warming and to allow easy access to samples
  • In addition to the above features the MiniSpin plus has:
  • Automatic rpm/rcf conversion
  • Extended time to 99 min or continous centrifugation
Everyday centrifugation tasks become easier with the MiniSpin® and MiniSpin® plus mini-centrifuges. Both offer soft-touch one-finger lid closure for ergonomic lid locking and automatic lid opening at the end of the run to prevent sample warming and to enable easy access to samples. Two models are available to meet your performance requirements: The centrifuge MiniSpin model delivers speeds up to 12,100×g and the MiniSpin plus model provides speed sufficient for molecular biology applications (up to 14,100×g).
Certifications: Certificate of quality/conformity (ISO 9001, ISO 13485, ISO 14001).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty