Would you like to log in again?
SP Bel-Art Sterileware® Large Sampling Spoon, Bel-Art Products, a part of SP
Supplier: Bel-Art
Sterileware®
H36948-0000
47727-682CS 98.97 USD InStock
47727-682
SP Bel-Art Sterileware® Large Sampling Spoon, Bel-Art Products, a part of SP
Spoons
Large, sterile sampling spoon is ideal for sampling semi-solids, viscous liquids, or bulk materials.
  • Sterile
  • Polystyrene plastic construction
  • Long handle protects against sample contact
  • Individually sealed in polyethylene bag
  • Lot stamped for accurate lot tracing
These 30 mL (1oz.) spoons are made from FDA-grade, high-impact polystyrene plastic
Individually sealed and sterilized in a polyethylene bag and ready for one-time use and disposal, they are lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization.
The long handle keeps the hand away from the sample.
Ordering information: Spoons are individually wrapped.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty