Would you like to log in again?
SP Bel-Art Stainless Steel Spoons, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
SCIENCEWARE®
H36729-0030 F36730-0015 H36729-0021 F36730-0030 F36730-0021 H36729-0015 H36729-0025 H36729-0018
47727-684PK 131.84 USD InStock
47727-684 13197-378 47727-690 13197-386 13197-384 13197-382 47727-686 13197-380
SP Bel-Art Stainless Steel Spoons, Bel-Art Products, a part of SP
Spoons
These double-ended, 304 stainless steel sampling spoons have a solid, rounded shaft.
  • Double-ended design
  • Stainless steel construction
  • Optional FEP coating provides non-stick surface
Available in plain polished stainless steel or coated with Fluo-Kem® Teflon® FEP, which complies with FDA regulations governing direct food contact applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty