Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Culture Tubes, Disposable, Borosilicate Glass

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
17729.576 27729.572 27729.584 27729.566 27729.574 27729.578 27729.580 27729.568 27729.583 27729.570 27729.586
47729-580CS 333.89 USD InStock
47729-580 47729-570 47729-572 47729-583 47729-584 47729-574 47729-586 47729-576 47729-566 47729-578 47729-568
VWR® Culture Tubes, Disposable, Borosilicate Glass
Tubes Culture Tubes
VWR® Disposable culture tubes are resistant to chemical attack and weathering.

  • Chemically inert
  • High heat tolerance
  • Uniform dimensions
  • Convenient dispensing packages

Manufactured from Type I, Class B borosilicate glass, each tube is heat-treated at temperatures exceeding 500 °C (932 °F) immediately before packing, which eliminates contamination, increases strength, and reduces breakage.

Contents will not leach out and contaminate solutions. Borosilicate glass construction prevents pH change in laboratory tests.

Tubes exceed USP Type I specifications. Tubes feature well-rounded bottoms and fire-polished tops free of rough edges. Dimensional specifications of these tubes are rigidly controlled for reliable, uniform results. Tubes are clean-sealed in a convenient dispensing package to keep them free from dust and other particulate contamination. Tube 47729-566 is also known as a Durham tube.

Please note: The capacity given below indicates the brim capacity (overflow capacity).

Ordering information: Due to machine packaging, quantities per case are approximate.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty