Would you like to log in again?
VWR® 12" Test Sieves, Half Height, Brass and Stainless Steel
Supplier: VWR
5/16BS12H 18BS12H 1/2"BS12H 70BS12H 3.5BS12H 60BS12H 20BS12H 100BS12H 1.00"BS12H 30BS12H 40BS12H 270BS12H 1/4"BS12H 12BS12H 4BS12H 230BS12H 500BS12H 120BS12H 8BS12H 325BS12H 5BS12H 635BS12H 50BS12H 1-1/2"BS12H 25BS12H .265BS12H 5/8"BS12H 1-1/4"BS12H 80BS12H 14BS12H 170BS12H 200BS12H 1/8BS12H 2"BS12H 7/16BS12H 10BS12H 16BS12H 450BS12H 6BS12H 400BS12H 3/8"BS12H 140BS12H 7BS12H 3/4"BS12H
47729-968EA 331.71 USD InStock
47729-968 47729-924 47729-946 47729-928 47729-908 47729-982 47729-960 47729-984 47729-964 47729-986 47729-988 47729-922 10811-524 10811-788 10811-522 10811-526 47729-990 10811-806 47729-978 47729-912 47729-914 47729-938 47729-918 47730-004 10811-780 47729-992 10811-460 47729-972 47729-994 47729-996 47729-952 10811-520 47729-998 10811-782 47729-976 47729-954 47729-932 10811-458 10811-612 10811-798 10811-610 10811-616 10811-614 47729-980
VWR® 12" Test Sieves, Half Height, Brass and Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Half Height (15/8") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Brass and stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.
Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.
Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty