Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® 12" Test Sieves, Half Height, Brass and Stainless Steel

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

5/16BS12H 18BS12H 1/2"BS12H 70BS12H 3.5BS12H 60BS12H 20BS12H 100BS12H 1.00"BS12H 30BS12H 40BS12H 270BS12H 1/4"BS12H 12BS12H 4BS12H 230BS12H 500BS12H 120BS12H 8BS12H 325BS12H 5BS12H 635BS12H 50BS12H 1-1/2"BS12H 25BS12H .265BS12H 5/8"BS12H 1-1/4"BS12H 80BS12H 14BS12H 170BS12H 200BS12H 1/8BS12H 2"BS12H 7/16BS12H 10BS12H 16BS12H 450BS12H 6BS12H 400BS12H 3/8"BS12H 140BS12H 7BS12H 3/4"BS12H
47729-968EA 331.71 USD InStock
47729-968 47729-924 47729-946 47729-928 47729-908 47729-982 47729-960 47729-984 47729-964 47729-986 47729-988 47729-922 10811-524 10811-788 10811-522 10811-526 47729-990 10811-806 47729-978 47729-912 47729-914 47729-938 47729-918 47730-004 10811-780 47729-992 10811-460 47729-972 47729-994 47729-996 47729-952 10811-520 47729-998 10811-782 47729-976 47729-954 47729-932 10811-458 10811-612 10811-798 10811-610 10811-616 10811-614 47729-980
VWR® 12" Test Sieves, Half Height, Brass and Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a larger sample size is required.

  • Half Height (15/8") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Brass and stainless steel construction

Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.

Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.

Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty