Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® 12" Test Sieves, Full Height, Brass and Stainless Steel

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

8BS12F 18BS12F 450BS12F 1/2"BS12F 12BS12F 7BS12F 500BS12F 60BS12F 80BS12F 400BS12F 35BS12F .265BS12F 5BS12F 7/16BS12F 14BS12F 3.5BS12F 16BS12F 3/8"BS12F 1/4BS12F 20BS12F 40BS12F 230BS12F 30BS12F 170BS12F 140BS12F 25BS12F 70BS12F 45BS12F 4BS12F 6BS12F 200BS12F 120BS12F 270BS12F 325BS12F 100BS12F 5/16"BS12F 635BS12F 10BS12F
47730-218EA 416.25 USD InStock
47730-218 47730-216 47730-214 47730-156 47730-212 47730-234 47730-198 47730-232 47730-210 47730-196 47730-174 47730-230 47730-194 10811-546 47730-192 47730-170 47730-190 10811-548 47730-208 47730-206 47730-228 47730-226 47730-202 47730-224 47730-200 47730-222 47730-168 47730-220 47730-188 10811-542 10811-540 47730-184 47730-162 47730-160 47730-182 47730-180 10811-538 10811-536
VWR® 12" Test Sieves, Full Height, Brass and Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a larger sample size is required.

  • Full Height (31/4") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Brass and stainless steel construction

Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.

Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.

Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty