Would you like to log in again?
VWR® 12" Test Sieves, Full Height, Brass and Stainless Steel
Supplier: VWR
8BS12F 18BS12F 450BS12F 1/2"BS12F 12BS12F 7BS12F 500BS12F 60BS12F 80BS12F 400BS12F 35BS12F .265BS12F 5BS12F 7/16BS12F 14BS12F 3.5BS12F 16BS12F 3/8"BS12F 1/4BS12F 20BS12F 40BS12F 230BS12F 30BS12F 170BS12F 140BS12F 25BS12F 70BS12F 45BS12F 4BS12F 6BS12F 200BS12F 120BS12F 270BS12F 325BS12F 100BS12F 5/16"BS12F 635BS12F 10BS12F
47730-218EA 416.25 USD InStock
47730-218 47730-216 47730-214 47730-156 47730-212 47730-234 47730-198 47730-232 47730-210 47730-196 47730-174 47730-230 47730-194 10811-546 47730-192 47730-170 47730-190 10811-548 47730-208 47730-206 47730-228 47730-226 47730-202 47730-224 47730-200 47730-222 47730-168 47730-220 47730-188 10811-542 10811-540 47730-184 47730-162 47730-160 47730-182 47730-180 10811-538 10811-536
VWR® 12" Test Sieves, Full Height, Brass and Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Full Height (31/4") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Brass and stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.
Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.
Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty