Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® 12" Test Sieves, Intermediate Height, All Stainless Steel

Supplier: VWR
1/4"SS12I 60SS12I 230SS12I 20SS12I 80SS12I 8SS12I
47730-438EA 449.22 USD InStock
47730-438 47730-434 47730-420 47730-394 47730-450 47730-408
VWR® 12" Test Sieves, Intermediate Height, All Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm ) all stainless steel test sieves are durable and ideal for sieving applications where a larger sample size is required.

  • Intermediate Height (2") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Stainless steel construction

Each sieve bears a traceable serial number and comes with a certificate of compliance verifying the sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes, and Centerline® sieve inspection or calibration certificates are available as accessories.

Certifications: Each serialized sieves comes with a dated ASTM E11 Certificate of Compliance.

Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.

Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth. Sieve Certification, Pans, and Covers must be ordered separately.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty