Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® 12" Test Sieves, Full Height, All Stainless Steel
Supplier: Avantor
18SS12F 4SS12F 1/4"SS12F 58SS12F 716SS12F 7SS12F 635SS12F 80SS12F 200SS12F 400SS12F 1.00"SS12F 20SS12F 50SS12F 325SS12F 60SS12F 516SS12F 170SS12F 140SS12F 5SS12F 70SS12F 120SS12F 230SS12F 35SS12F 25SS12F 100SS12F 38SS12F 12SS12F 34SS12F 450SS12F 500SS12F 14SS12F 78SS12F 8SS12F 3.5SS12F .530SS12F 45SS12F 12SS12F 270SS12F 10SS12F 16SS12F 30SS12F .265"SS12F 40SS12F 6SS12F
47730-538EA 460.94 USD InStock
47730-538 47730-516 47730-514 47730-536 47730-512 47730-534 47730-556 47730-532 47730-554 47730-552 47730-574 47730-530 10811-592 47730-550 47730-570 10811-596 10811-594 10811-600 10811-786 10811-726 10811-604 10811-602 10811-608 10811-728 10811-606 47730-508 47730-528 47730-506 47730-548 47730-526 47730-546 47730-568 47730-524 47730-488 47730-522 47730-544 47730-520 47730-564 47730-542 47730-562 47730-540 10811-732 10811-730 10811-598
VWR® 12" Test Sieves, Full Height, All Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm ) all stainless steel test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Full Height (31/4") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying the sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes, and Centerline® sieve inspection or calibration certificates are available as accessories.
Certifications: Each serialized sieves comes with a dated ASTM E11 Certificate of Compliance.
Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.
Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth. Sieve Certification, Pans, and Covers must be ordered separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty