Would you like to log in again?
VWR® 12" Test Sieves, Full Height, All Stainless Steel
Supplier: VWR
18SS12F 4SS12F 1/4"SS12F 5/8SS12F 7/16SS12F 7SS12F 635SS12F 80SS12F 200SS12F 400SS12F 1.00"SS12F 20SS12F 50SS12F 325SS12F 60SS12F 5/16SS12F 170SS12F 140SS12F 5SS12F 70SS12F 120SS12F 230SS12F 35SS12F 25SS12F 100SS12F 3/8SS12F 1/2SS12F 3/4SS12F 450SS12F 500SS12F 14SS12F 7/8SS12F 8SS12F 3.5SS12F .530SS12F 45SS12F 12SS12F 270SS12F 10SS12F 16SS12F 30SS12F .265"SS12F 40SS12F 6SS12F
47730-538EA 460.94 USD InStock
47730-538 47730-516 47730-514 47730-536 47730-512 47730-534 47730-556 47730-532 47730-554 47730-552 47730-574 47730-530 10811-592 47730-550 47730-570 10811-596 10811-594 10811-600 10811-786 10811-726 10811-604 10811-602 10811-608 10811-728 10811-606 47730-508 47730-528 47730-506 47730-548 47730-526 47730-546 47730-568 47730-524 47730-488 47730-522 47730-544 47730-520 47730-564 47730-542 47730-562 47730-540 10811-732 10811-730 10811-598
VWR® 12" Test Sieves, Full Height, All Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm ) all stainless steel test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Full Height (31/4") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying the sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes, and Centerline® sieve inspection or calibration certificates are available as accessories.
Certifications: Each serialized sieves comes with a dated ASTM E11 Certificate of Compliance.
Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.
Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth. Sieve Certification, Pans, and Covers must be ordered separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty